ENA444722996 Enamine

6-bromophthalazin-1-ol

  • MDL number MFCD09264003
  • Cas Number : 75884-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-192438
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart