ENA444723004 Enamine

5-(piperidin-1-yl)quinolin-8-amine

  • MDL number MFCD12412178
  • Cas Number : 1152988-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192884
Availability details for
Quantity
  • Availability
  • Add to Cart