ENA444723007 Enamine

2-(methoxymethyl)-5-phenyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD21429745
  • Cas Number : 1480580-45-7
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-193122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193122
Availability details for
Quantity
  • Availability
  • Add to Cart