ENA444723013 Enamine

3,3-dimethyl-5-oxo-5-phenylpentanoic acid

  • MDL number MFCD17253588
  • Cas Number : 20633-28-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-193341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193341
Availability details for
Quantity
  • Availability
  • Add to Cart