ENA444723023 Enamine

1-(4-fluorophenyl)-6-methoxy-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD18788400
  • Cas Number : 1291846-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-193708
Availability details for
Quantity
  • Availability
  • Add to Cart