ENA444723037 Enamine

1-(trifluoroacetyl)azetidin-3-one

  • MDL number MFCD28246252
  • Cas Number : 1803610-57-2
  • Molecular Weight : 167.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-194535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194535
Availability details for
Quantity
  • Availability
  • Add to Cart