ENA444723051 Enamine

4-bromo-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD22378818
  • Cas Number : 1008769-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-195058
Availability details for
Quantity
  • Availability
  • Add to Cart