ENA444723073 Enamine

6-bromo-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD12068434
  • Cas Number : 99853-21-1
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-195516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195516
Availability details for
Quantity
  • Availability
  • Add to Cart