ENA444723112 Enamine

(carbamothioylamino)urea

  • MDL number MFCD00031451
  • Cas Number : 2937-80-6
  • Molecular Weight : 134.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-216
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196191

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-196191
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart