ENA444723127 Enamine

methyl 2-chloro-6-methyl-3-nitropyridine-4-carboxylate

  • MDL number MFCD28348044
  • Cas Number : 1465253-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-196466
Availability details for
Quantity
  • Availability
  • Add to Cart