ENA444723149 Enamine

[2-(methoxymethyl)-5-phenyl-1,3-thiazol-4-yl]methanamine

  • MDL number MFCD20402085
  • Cas Number : 1340521-16-5
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196730
Availability details for
Quantity
  • Availability
  • Add to Cart