ENA444723170 Enamine

methyl 1,3-dimethyl-4-nitro-1H-pyrazole-5-carboxylate

  • MDL number MFCD00662662
  • Cas Number : 78208-69-2
  • Molecular Weight : 199.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197243
Availability details for
Quantity
  • Availability
  • Add to Cart