ENA444723208 Enamine

tert-butyl N-[2-(1-hydroxyethyl)-3-methoxyphenyl]carbamate

  • MDL number MFCD24475040
  • Cas Number : 380225-70-7
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197706
Availability details for
Quantity
  • Availability
  • Add to Cart