ENA444723219 Enamine

5-bromo-2-cyanobenzene-1-sulfonyl chloride

  • MDL number MFCD18390734
  • Cas Number : 1257415-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142594
Availability details for
Quantity
  • Availability
  • Add to Cart