ENA444723250 Enamine

5-(hydroxymethyl)-3-methylfuran-2-carboxylic acid

  • MDL number MFCD12093268
  • Cas Number : 121756-65-8
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-147850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147850
Availability details for
Quantity
  • Availability
  • Add to Cart