ENA444723257 Enamine

1-(5-tert-butylfuran-2-yl)ethan-1-one

  • MDL number MFCD24691838
  • Cas Number : 32180-98-6
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197921
Availability details for
Quantity
  • Availability
  • Add to Cart