ENA444723268 Enamine

2-(3-methyl-1,2,4-oxadiazol-5-yl)cyclopentan-1-one

  • MDL number MFCD18335253
  • Cas Number : 1343762-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-197952
Availability details for
Quantity
  • Availability
  • Add to Cart