ENA444723280 Enamine

2,4-dichloro-1-ethylbenzene

  • MDL number MFCD01657119
  • Cas Number : 54484-62-7
  • Molecular Weight : 175.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198007
Availability details for
Quantity
  • Availability
  • Add to Cart