ENA444723300 Enamine

(4-chlorophenyl)(oxan-4-yl)methanol

  • MDL number MFCD17243924
  • Cas Number : 1341482-24-3
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198097
Availability details for
Quantity
  • Availability
  • Add to Cart