ENA444723352 Enamine

ethyl 5-(propan-2-yl)-1,3,4-thiadiazole-2-carboxylate

  • MDL number MFCD19981641
  • Cas Number : 1330754-00-1
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198283
Availability details for
Quantity
  • Availability
  • Add to Cart