ENA444723433 Enamine

3-amino-2-(cyclopentylmethyl)propan-1-ol

  • MDL number MFCD16706732
  • Cas Number : 1248717-01-2
  • Molecular Weight : 157.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198518
Availability details for
Quantity
  • Availability
  • Add to Cart