ENA444723445 Enamine

{1,5-diazabicyclo[3.2.2]nonan-6-yl}methanamine

  • MDL number MFCD28348011
  • Cas Number : 1507918-33-3
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170678
Availability details for
Quantity
  • Availability
  • Add to Cart