ENA444723481 Enamine

N,N-dimethyl-5-(piperidin-4-yl)-1H-1,2,4-triazol-3-amine dihydrochloride

  • MDL number MFCD28246373
  • Cas Number : 1803582-03-7
  • Molecular Weight : 268.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198899
Availability details for
Quantity
  • Availability
  • Add to Cart