ENA444723564 Enamine

1-(4-chloro-3-fluorophenyl)ethane-1,2-diol

  • MDL number MFCD24287488
  • Cas Number : 1558116-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-199415
Availability details for
Quantity
  • Availability
  • Add to Cart