ENA444723589 Enamine

1-(4-fluorophenyl)-4-methoxybutan-1-one

  • MDL number MFCD12058711
  • Cas Number : 71434-08-7
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199519
Availability details for
Quantity
  • Availability
  • Add to Cart