ENA444723604 Enamine

2-hydroxy-1-(morpholin-4-yl)propan-1-one

  • MDL number MFCD12137913
  • Cas Number : 27097-66-1
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199538
Availability details for
Quantity
  • Availability
  • Add to Cart