ENA444723620 Enamine

2,4-difluorophenyl chloroformate

  • MDL number MFCD24141121
  • Cas Number : 400900-00-7
  • Molecular Weight : 192.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199635

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P311,P312,P330,P361,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-199635
pictograms GHS05,GHS06
hstatements H227,H301,H311,H314,H331
Availability details for
Quantity
  • Availability
  • Add to Cart