ENA444723663 Enamine

2-(4-methyl-1,3-benzoxazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28246408
  • Cas Number : 1803583-70-1
  • Molecular Weight : 249.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199735
Availability details for
Quantity
  • Availability
  • Add to Cart