ENA444723681 Enamine

N-(2-aminoethyl)-4-chloro-2-fluorobenzene-1-sulfonamide hydrochloride

  • MDL number MFCD28246418
  • Cas Number : 1568443-45-7
  • Molecular Weight : 289.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199760
Availability details for
Quantity
  • Availability
  • Add to Cart