ENA444723923 Enamine

(3-amino-4-methylphenyl)(pyridin-2-yl)methanol

  • MDL number MFCD28348059
  • Cas Number : 1803581-37-4
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199944
Availability details for
Quantity
  • Availability
  • Add to Cart