ENA444723928 Enamine

2-(5-methyl-1H-1,2,4-triazol-3-yl)piperidine hydrochloride

  • MDL number MFCD28246434
  • Cas Number : 1803593-85-2
  • Molecular Weight : 202.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200028
Availability details for
Quantity
  • Availability
  • Add to Cart