ENA444723959 Enamine

N-methyl-N-[(pyrimidin-2-yl)methyl]pyrrolidin-3-amine trihydrochloride

  • MDL number MFCD28246479
  • Cas Number : 1803581-51-2
  • Molecular Weight : 301.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200593
Availability details for
Quantity
  • Availability
  • Add to Cart