ENA444723961 Enamine

5-(chloromethyl)-1-(2-fluoroethyl)-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD28246482
  • Cas Number : 1803592-97-3
  • Molecular Weight : 200.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200608
Availability details for
Quantity
  • Availability
  • Add to Cart