ENA444723965 Enamine

(2-tert-butyl-1,3-oxazol-4-yl)methanol

  • MDL number MFCD28348068
  • Cas Number : 1448866-07-6
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200677
Availability details for
Quantity
  • Availability
  • Add to Cart