ENA444723967 Enamine

3-amino-2-chloro-6-methylpyridine-4-carboxylic acid hydrochloride

  • MDL number MFCD28246487
  • Cas Number : 1803610-45-8
  • Molecular Weight : 223.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200688
Availability details for
Quantity
  • Availability
  • Add to Cart