ENA444723971 Enamine

tert-butyl 6-methyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate

  • MDL number MFCD28246493
  • Cas Number : 1803594-40-2
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200792
Availability details for
Quantity
  • Availability
  • Add to Cart