ENA444723982 Enamine

tert-butyl N-{[3-(3,4-dimethylphenyl)-4,5-dihydro-1,2-oxazol-5-yl]methyl}carbamate

  • MDL number MFCD28348077
  • Cas Number : 1803612-22-7
  • Molecular Weight : 304.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200854
Availability details for
Quantity
  • Availability
  • Add to Cart