ENA444723994 Enamine

(2R)-1-{[1-(4-chlorophenyl)propan-2-yl]amino}propan-2-ol hydrochloride

  • MDL number MFCD28246509
  • Cas Number : 1807937-65-0
  • Molecular Weight : 264.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200922
Availability details for
Quantity
  • Availability
  • Add to Cart