ENA444724063 Enamine

3-[(4-chlorophenyl)methyl]oxolan-3-amine hydrochloride

  • MDL number MFCD28246583
  • Cas Number : 1803585-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201745
Availability details for
Quantity
  • Availability
  • Add to Cart