ENA444724075 Enamine

[(3-aminopropyl)sulfanyl]carbonitrile hydrobromide

  • MDL number MFCD28348093
  • Cas Number : 1803600-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201896
Availability details for
Quantity
  • Availability
  • Add to Cart