ENA444724082 Enamine

3-fluoro-3-(2-fluorophenyl)pyrrolidine hydrochloride

  • MDL number MFCD28246597
  • Cas Number : 1803561-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201971
Availability details for
Quantity
  • Availability
  • Add to Cart