ENA444724094 Enamine

methyl 2-[(2-methoxy-2-oxoethyl)sulfanyl]-2-methylpropanoate

  • MDL number MFCD28348105
  • Cas Number : 1803561-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202194
Availability details for
Quantity
  • Availability
  • Add to Cart