ENA444724100 Enamine

1-cyclopropylpiperazin-2-one hydrochloride

  • MDL number MFCD28348111
  • Cas Number : 1803584-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202232
Availability details for
Quantity
  • Availability
  • Add to Cart