ENA444724103 Enamine

1-[2-(4-fluorophenoxy)ethyl]-5-methyl-1H-1,2,3-triazol-4-amine hydrochloride

  • MDL number MFCD28348115
  • Cas Number : 1803595-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202263
Availability details for
Quantity
  • Availability
  • Add to Cart