ENA444724104 Enamine

sodium 5-(propan-2-yl)-1,3,4-thiadiazole-2-carboxylate

  • MDL number MFCD28348116
  • Cas Number : 1803586-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202265
Availability details for
Quantity
  • Availability
  • Add to Cart