ENA444724106 Enamine

1-[1-methyl-3,5-bis(trifluoromethyl)-1H-pyrrol-2-yl]ethan-1-one

  • MDL number MFCD28348120
  • Cas Number : 1803611-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202285
Availability details for
Quantity
  • Availability
  • Add to Cart