ENA444724111 Enamine

1-[3-(4-methoxyphenyl)-4,5-dihydro-1,2-oxazol-5-yl]methanamine hydrochloride

  • MDL number MFCD28348126
  • Cas Number : 1803589-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202341
Availability details for
Quantity
  • Availability
  • Add to Cart