ENA444724114 Enamine

1-[3-(4-methoxy-3-methylphenyl)-4,5-dihydro-1,2-oxazol-5-yl]methanamine hydrochloride

  • MDL number MFCD28348129
  • Cas Number : 1803600-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202349
Availability details for
Quantity
  • Availability
  • Add to Cart