ENA444724115 Enamine

3-fluoro-3-phenylpyrrolidine hydrochloride

  • MDL number MFCD28348130
  • Cas Number : 1803603-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202352
Availability details for
Quantity
  • Availability
  • Add to Cart