ENA444724118 Enamine

2-(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28348133
  • Cas Number : 1803600-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202405
Availability details for
Quantity
  • Availability
  • Add to Cart